Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGCTGCTGTGTTGCCTCCCCA[A/G]CTCCCCCAGCCCCCGGGCCCGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 106195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC4A3 - solute carrier family 4 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326559.1 | 1092 | Missense Mutation | AAC,AGC | N,S 231 | NP_001313488.1 | |
NM_005070.3 | 1092 | Intron | NP_005061.2 | |||
NM_201574.2 | 1092 | Missense Mutation | AAC,AGC | N,S 231 | NP_963868.2 | |
XM_005246789.4 | 1092 | Missense Mutation | AAC,AGC | N,S 231 | XP_005246846.1 | |
XM_005246790.4 | 1092 | Intron | XP_005246847.1 | |||
XM_011511665.2 | 1092 | Missense Mutation | AAC,AGC | N,S 178 | XP_011509967.1 | |
XM_011511667.2 | 1092 | Intron | XP_011509969.1 |