Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACCTCCTCACTTGTGGCACCTT[C/T]GCTTTTGATCCGAAGTGCGGGGTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603706 | ||||||||||||||||||||
Literature Links: |
M1AP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
M1AP - meiosis 1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4F - ssemaphorin 4F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271661.1 | 578 | Intron | NP_001258590.1 | |||
NM_001271662.1 | 578 | Intron | NP_001258591.1 | |||
NM_004263.4 | 578 | Silent Mutation | TTC,TTT | F,F 143 | NP_004254.2 | |
XM_011532475.2 | 578 | Missense Mutation | TTC,TTT | F,F 143 | XP_011530777.1 | |
XM_011532476.2 | 578 | Missense Mutation | TTC,TTT | F,F 143 | XP_011530778.1 | |
XM_011532477.2 | 578 | Missense Mutation | TTC,TTT | F,F 143 | XP_011530779.1 | |
XM_017003172.1 | 578 | Missense Mutation | TTC,TTT | F,F 41 | XP_016858661.1 | |
XM_017003173.1 | 578 | Intron | XP_016858662.1 | |||
XM_017003174.1 | 578 | Intron | XP_016858663.1 |