Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGGTTGACGACGATTTCCTTGC[C/T]GTTCTCGGTGCCATCACCGAACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT14 - polypeptide N-acetylgalactosaminyltransferase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253826.1 | 1758 | Missense Mutation | AGC,GGC | S,G 535 | NP_001240755.1 | |
NM_001253827.1 | 1758 | Missense Mutation | AGC,GGC | S,G 510 | NP_001240756.1 | |
NM_024572.3 | 1758 | Missense Mutation | AGC,GGC | S,G 530 | NP_078848.2 | |
XM_011533105.1 | 1758 | Missense Mutation | AGC,GGC | S,G 510 | XP_011531407.1 | |
XM_017004906.1 | 1758 | Missense Mutation | AGC,GGC | S,G 541 | XP_016860395.1 | |
XM_017004907.1 | 1758 | UTR 3 | XP_016860396.1 | |||
XM_017004908.1 | 1758 | UTR 3 | XP_016860397.1 |