Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACATGGTATGAGCCTTCATCCC[A/G]GTTTCTGTACTTGTACATGGCATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600565 | ||||||||||||||||||||
Literature Links: |
NRXN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRXN1 - neurexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264642.3 | 5828 | Missense Mutation | CGG,TGG | R,W 1462 | XP_005264699.1 | |
XM_005264643.3 | 5828 | Missense Mutation | CGG,TGG | R,W 1447 | XP_005264700.1 | |
XM_006712137.3 | 5828 | Missense Mutation | CGG,TGG | R,W 1432 | XP_006712200.1 | |
XM_006712140.3 | 5828 | Intron | XP_006712203.2 | |||
XM_011533167.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1465 | XP_011531469.1 | |
XM_011533171.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1458 | XP_011531473.1 | |
XM_011533172.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1456 | XP_011531474.1 | |
XM_011533174.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1450 | XP_011531476.1 | |
XM_011533175.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1446 | XP_011531477.1 | |
XM_011533177.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1435 | XP_011531479.1 | |
XM_011533178.2 | 5828 | Missense Mutation | CGG,TGG | R,W 1435 | XP_011531480.1 | |
XM_011533180.2 | 5828 | Intron | XP_011531482.1 | |||
XM_011533183.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1176 | XP_011531485.1 | |
XM_017005303.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1474 | XP_016860792.1 | |
XM_017005304.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1473 | XP_016860793.1 | |
XM_017005305.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1471 | XP_016860794.1 | |
XM_017005306.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1470 | XP_016860795.1 | |
XM_017005307.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1468 | XP_016860796.1 | |
XM_017005308.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1467 | XP_016860797.1 | |
XM_017005309.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1465 | XP_016860798.1 | |
XM_017005310.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1464 | XP_016860799.1 | |
XM_017005311.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1459 | XP_016860800.1 | |
XM_017005312.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1457 | XP_016860801.1 | |
XM_017005313.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1455 | XP_016860802.1 | |
XM_017005314.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1454 | XP_016860803.1 | |
XM_017005315.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1453 | XP_016860804.1 | |
XM_017005316.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1453 | XP_016860805.1 | |
XM_017005317.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1451 | XP_016860806.1 | |
XM_017005318.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1450 | XP_016860807.1 | |
XM_017005319.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1449 | XP_016860808.1 | |
XM_017005320.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1446 | XP_016860809.1 | |
XM_017005321.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1444 | XP_016860810.1 | |
XM_017005322.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1441 | XP_016860811.1 | |
XM_017005323.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1436 | XP_016860812.1 | |
XM_017005324.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1426 | XP_016860813.1 | |
XM_017005325.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1423 | XP_016860814.1 | |
XM_017005326.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1422 | XP_016860815.1 | |
XM_017005327.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1417 | XP_016860816.1 | |
XM_017005328.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1411 | XP_016860817.1 | |
XM_017005329.1 | 5828 | Intron | XP_016860818.1 | |||
XM_017005330.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1403 | XP_016860819.1 | |
XM_017005331.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1398 | XP_016860820.1 | |
XM_017005332.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1391 | XP_016860821.1 | |
XM_017005333.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1388 | XP_016860822.1 | |
XM_017005334.1 | 5828 | Missense Mutation | CGG,TGG | R,W 1154 | XP_016860823.1 | |
XM_017005335.1 | 5828 | Missense Mutation | CGG,TGG | R,W 423 | XP_016860824.1 | |
XM_017005336.1 | 5828 | Missense Mutation | CGG,TGG | R,W 420 | XP_016860825.1 | |
XM_017005337.1 | 5828 | Missense Mutation | CGG,TGG | R,W 390 | XP_016860826.1 |