Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTTCTATGTTCAGGATATTCTT[C/G]AAAGTCCTAAAAATAAGTGGTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602559 | ||||||||||||||||||||
Literature Links: |
XPO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XPO1 - exportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003400.3 | 3243 | Missense Mutation | CAA,GAA | Q,E 839 | NP_003391.1 | |
XM_005264544.1 | 3243 | Missense Mutation | CAA,GAA | Q,E 824 | XP_005264601.1 | |
XM_005264546.1 | 3243 | Missense Mutation | CAA,GAA | Q,E 708 | XP_005264603.1 | |
XM_006712094.2 | 3243 | Missense Mutation | CAA,GAA | Q,E 839 | XP_006712157.1 | |
XM_011533097.1 | 3243 | Missense Mutation | CAA,GAA | Q,E 839 | XP_011531399.1 | |
XM_011533098.1 | 3243 | Missense Mutation | CAA,GAA | Q,E 794 | XP_011531400.1 | |
XM_011533099.2 | 3243 | Missense Mutation | CAA,GAA | Q,E 773 | XP_011531401.1 |