Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCCAGGGAGGAGGCGGCTGAGA[C/T]CGGCCCGCGGTTCGTGGCCCTGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613196 | ||||||||||||||||||||
Literature Links: |
ETAA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETAA1 - Ewing tumor associated antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019002.3 | 247 | Missense Mutation | CCG,TCG | P,S 39 | NP_061875.2 | |
XM_017004376.1 | 247 | Missense Mutation | CCG,TCG | P,S 39 | XP_016859865.1 | |
XM_017004377.1 | 247 | Missense Mutation | CCG,TCG | P,S 39 | XP_016859866.1 |