Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCAGGAAGGACTGGGATCTGTC[A/G]CTGGCGACTCCCAAAGGTATGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM131 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM131 - transmembrane protein 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015348.1 | 5712 | Missense Mutation | GCG,GTG | A,V 1770 | NP_056163.1 | |
XM_005263909.1 | 5712 | Missense Mutation | GCG,GTG | A,V 1787 | XP_005263966.1 | |
XM_005263910.1 | 5712 | Missense Mutation | GCG,GTG | A,V 1786 | XP_005263967.1 | |
XM_005263911.1 | 5712 | Missense Mutation | GCG,GTG | A,V 1748 | XP_005263968.1 | |
XM_005263912.3 | 5712 | Missense Mutation | GCG,GTG | A,V 1731 | XP_005263969.1 | |
XM_011510869.1 | 5712 | Missense Mutation | GCG,GTG | A,V 1725 | XP_011509171.1 |