Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCTGGCAGGTACATGTTGATCA[A/G]CAGGGGCTGGGAGGCGCCGCTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600836 MIM: 607150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYBA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYBA2 - crystallin beta A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FEV - FEV, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017521.2 | 613 | Silent Mutation | CTG,TTG | L,L 11 | NP_059991.1 |
LINC00608 - long intergenic non-protein coding RNA 608 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |