Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAGTTCGACGAGCAAGCTTAAA[A/G]AACTCTTCTGCAGCATCAAGTGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609207 | ||||||||||||||||||||
Literature Links: |
MREG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MREG - melanoregulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018000.2 | 842 | Silent Mutation | NP_060470.2 | |||
XM_011511468.2 | 842 | Silent Mutation | XP_011509770.1 | |||
XM_017004475.1 | 842 | Intron | XP_016859964.1 |