Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCTGGAGAGAGTTTCGGCTGA[A/G]GCCTCTGCAGTTGGATGGCAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf83 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf83 - chromosome 2 open reading frame 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162483.1 | 645 | UTR 3 | NP_001155955.1 | |||
NM_020161.3 | 645 | Intron | NP_064546.3 |
LOC729968 - spore coat protein SP96 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |