Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGCGCTGCTTTACCAGGGCCCC[A/C]ACAACACGCTGTTCGGCTACTCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGA4 - integrin subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000885.5 | 905 | Missense Mutation | AAC,CAC | N,H 48 | NP_000876.3 | |
NM_001316312.1 | 905 | Missense Mutation | AAC,CAC | N,H 48 | NP_001303241.1 |