Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAAAATGGAGGTAAAACCGCCG[C/T]CCGGTCGCCCCCAGCCCGACTCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605372 | ||||||||||||||||||||
Literature Links: |
HNRNPA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPA3 - heterogeneous nuclear ribonucleoprotein A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246380.1 | 125 | Missense Mutation | CCC,TCC | P,S 7 | XP_005246437.1 | |
XM_005246381.4 | 125 | Intron | XP_005246438.1 | |||
XM_006712365.2 | 125 | Missense Mutation | CCC,TCC | P,S 7 | XP_006712428.1 | |
XM_017003587.1 | 125 | Intron | XP_016859076.1 | |||
XM_017003588.1 | 125 | Missense Mutation | CCC,TCC | P,S 7 | XP_016859077.1 |
LOC105373759 - uncharacterized LOC105373759 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017005483.1 | 125 | Intron | XP_016860972.1 |
MIR4444-1 - microRNA 4444-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |