Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGAAGGAAAAATCATATTTTTG[C/T]GTGATGTCATAGATCCACCCTACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC16A14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC16A14 - solute carrier family 16 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152527.4 | 1716 | Silent Mutation | ACA,ACG | T,T 467 | NP_689740.2 | |
XM_005246353.3 | 1716 | Silent Mutation | ACA,ACG | T,T 419 | XP_005246410.1 | |
XM_011510751.2 | 1716 | Silent Mutation | ACA,ACG | T,T 467 | XP_011509053.1 | |
XM_011510752.1 | 1716 | Intron | XP_011509054.1 | |||
XM_017003480.1 | 1716 | Silent Mutation | ACA,ACG | T,T 467 | XP_016858969.1 |