Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTATTTCACTGGAAACCCCTTAT[G/T]TTGAGAGCTCCAGGAAGCGCCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142984 MIM: 142986 MIM: 142988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXD10 - homeobox D10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD11 - homeobox D11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021192.2 | 1013 | Missense Mutation | TGT,TTT | C,F 338 | NP_067015.2 |
HOXD12 - homeobox D12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985805 - sine oculis-binding protein homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |