Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATTATTTACTCAGAGTTCTTG[C/T]TGTAGGCTTTGTCTTGAATGTGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C2orf69 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf69 - chromosome 2 open reading frame 69 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYW5 - tRNA-yW synthesizing protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039693.2 | 1198 | Missense Mutation | AAC,AGC | N,S 311 | NP_001034782.1 | |
XM_017003310.1 | 1198 | Missense Mutation | AAC,AGC | N,S 206 | XP_016858799.1 |