Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGCTACTTAAGTCTCATCTCAC[C/T]ATTATAGACCATTTGAAAATTAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102581 MIM: 603056 | ||||||||||||||||||||
Literature Links: |
ACVR2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACVR2A - activin A receptor type 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORC4 - origin recognition complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190879.2 | 1333 | Missense Mutation | AAG,GAG | K,E 352 | NP_001177808.1 | |
NM_001190881.2 | 1333 | Intron | NP_001177810.1 | |||
NM_001190882.2 | 1333 | Missense Mutation | AAG,GAG | K,E 278 | NP_001177811.1 | |
NM_002552.4 | 1333 | Missense Mutation | AAG,GAG | K,E 352 | NP_002543.2 | |
NM_181741.3 | 1333 | Missense Mutation | AAG,GAG | K,E 352 | NP_859525.1 | |
NM_181742.3 | 1333 | Intron | NP_859526.1 | |||
XM_011511255.2 | 1333 | Missense Mutation | AAG,GAG | K,E 352 | XP_011509557.1 | |
XM_017004226.1 | 1333 | Missense Mutation | AAG,GAG | K,E 352 | XP_016859715.1 | |
XM_017004227.1 | 1333 | Missense Mutation | AAG,GAG | K,E 268 | XP_016859716.1 |