Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTGTAGCGGCGCGCCCGGAGGCT[A/C]CCCCAGCCAAAGAGTGCCCAGCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142991 MIM: 142988 MIM: 142989 | ||||||||||||||||||||
Literature Links: |
EVX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EVX2 - even-skipped homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD12 - homeobox D12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD13 - homeobox D13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000523.3 | 391 | Missense Mutation | ACC,CCC | T,P 102 | NP_000514.2 | |
XM_011511068.2 | 391 | Intron | XP_011509370.1 | |||
XM_011511069.2 | 391 | Intron | XP_011509371.1 |