Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTATCAAGTTTGTAATAAGAGTG[C/T]TGATCAAATTGGCACAGTTTGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615295 | ||||||||||||||||||||
Literature Links: |
AHSA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AHSA2 - AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321300.1 | 9798 | Intron | NP_001308229.1 | |||
NM_001321301.1 | 9798 | Intron | NP_001308230.1 | |||
NM_001321302.1 | 9798 | Intron | NP_001308231.1 | |||
NM_001321303.1 | 9798 | Intron | NP_001308232.1 | |||
NM_001321304.1 | 9798 | Intron | NP_001308233.1 | |||
NM_001321305.1 | 9798 | Intron | NP_001308234.1 | |||
NM_152392.4 | 9798 | Intron | NP_689605.1 | |||
XM_011532541.2 | 9798 | UTR 3 | XP_011530843.1 | |||
XM_011532543.2 | 9798 | UTR 3 | XP_011530845.1 | |||
XM_011532545.2 | 9798 | UTR 3 | XP_011530847.1 | |||
XM_017003377.1 | 9798 | UTR 3 | XP_016858866.1 | |||
XM_017003378.1 | 9798 | UTR 3 | XP_016858867.1 |
USP34 - ubiquitin specific peptidase 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014709.3 | 9798 | Missense Mutation | AAC,AGC | N,S 3259 | NP_055524.3 |