Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGGCATCTTGAACCTCTTGAT[G/T]TTCTTTATCAGCTTTGCATTTTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606093 | ||||||||||||||||||||
Literature Links: |
CCDC173 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC173 - coiled-coil domain containing 173 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085447.1 | 1297 | Missense Mutation | NP_001078916.1 | |||
XM_011510589.1 | 1297 | Missense Mutation | XP_011508891.1 | |||
XM_011510590.1 | 1297 | Missense Mutation | XP_011508892.1 |
PPIG - peptidylprolyl isomerase G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |