Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGGAGCCGGGTCAGGACGCTGC[A/G]CCAGGCCTTCTTGGCCTTGCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609635 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCF23 - transcription factor 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175769.2 | 335 | Missense Mutation | CAC,CGC | H,R 93 | NP_786951.1 | |
XM_005264159.4 | 335 | Missense Mutation | CAC,CGC | H,R 93 | XP_005264216.1 |