Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCTCAGGTCCTGGCTGCTTCG[A/G]ACATAGCCAGCACCCCCACTGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYG1 - lysozyme g1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174898.2 | 738 | Silent Mutation | GTC,GTT | V,V 168 | NP_777558.1 | |
XM_005263869.2 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_005263926.1 | |
XM_005263870.3 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_005263927.1 | |
XM_005263871.4 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_005263928.1 | |
XM_011510580.2 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_011508882.1 | |
XM_017003311.1 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_016858800.1 | |
XM_017003312.1 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_016858801.1 | |
XM_017003313.1 | 738 | Silent Mutation | GTC,GTT | V,V 168 | XP_016858802.1 |