Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGTGGTGCACTGTGTGCCCCA[C/T]GTGGGCCGAGGGCAGCGGCGGGTGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610166 | |||||||||||||||||||||||
Literature Links: |
IQSEC1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IQSEC1 - IQ motif and Sec7 domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134382.2 | 3691 | Missense Mutation | ATG,GTG | M,V 1084 | NP_001127854.1 | |
NM_014869.6 | 3691 | Intron | NP_055684.3 | |||
XM_011534304.2 | 3691 | Missense Mutation | ATG,GTG | M,V 1192 | XP_011532606.1 | |
XM_011534305.2 | 3691 | Missense Mutation | ATG,GTG | M,V 1102 | XP_011532607.1 | |
XM_011534306.2 | 3691 | Missense Mutation | ATG,GTG | M,V 1099 | XP_011532608.1 | |
XM_011534307.2 | 3691 | Missense Mutation | ATG,GTG | M,V 1099 | XP_011532609.1 | |
XM_011534308.2 | 3691 | Missense Mutation | ATG,GTG | M,V 1098 | XP_011532610.1 | |
XM_011534310.2 | 3691 | UTR 3 | XP_011532612.1 | |||
XM_011534311.2 | 3691 | Missense Mutation | ATG,GTG | M,V 976 | XP_011532613.1 | |
XM_011534312.2 | 3691 | Missense Mutation | ATG,GTG | M,V 976 | XP_011532614.1 | |
XM_011534313.2 | 3691 | Missense Mutation | ATG,GTG | M,V 976 | XP_011532615.1 | |
XM_011534314.2 | 3691 | Missense Mutation | ATG,GTG | M,V 976 | XP_011532616.1 | |
XM_011534315.2 | 3691 | UTR 3 | XP_011532617.1 | |||
XM_017007581.1 | 3691 | Missense Mutation | ATG,GTG | M,V 1182 | XP_016863070.1 | |
XM_017007582.1 | 3691 | Missense Mutation | ATG,GTG | M,V 1177 | XP_016863071.1 |