Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGGGCAGGTGCCGCACAGGAC[A/G]ACAGGTGTTTCTGGTACCCAGCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605474 MIM: 607433 | ||||||||||||||||||||
Literature Links: |
LOC101929054 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929054 - uncharacterized LOC101929054 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLR9 - toll like receptor 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TWF2 - twinfilin actin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007284.3 | 567 | Missense Mutation | TCG,TTG | S,L 139 | NP_009215.1 |