Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTATGAGGGCCTCCTTTAAGTCT[C/T]GGTTCTTTTGTTCTTTGAAGCGTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605931 | |||||||||||||||||||||||
Literature Links: |
SNX4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SNX4 - sorting nexin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003794.3 | 1377 | Missense Mutation | CAA,CGA | Q,R 416 | NP_003785.1 | |
XM_017007414.1 | 1377 | Missense Mutation | CAA,CGA | Q,R 448 | XP_016862903.1 |