Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCCTGAGCGGCGGCCTGGGCAG[G/T]TCCCCGTTCTTGCCGATGCCCATGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608922 MIM: 616588 | |||||||||||||||||||||||
Literature Links: |
ARL13B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARL13B - ADP ribosylation factor like GTPase 13B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DHFR2 - dihydrofolate reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195643.1 | 516 | Missense Mutation | GAA,GAC | E,D 22 | NP_001182572.1 | |
NM_176815.4 | 516 | Missense Mutation | GAA,GAC | E,D 22 | NP_789785.1 | |
XM_011512537.2 | 516 | Missense Mutation | GAA,GAC | E,D 22 | XP_011510839.1 |
NSUN3 - NOP2/Sun RNA methyltransferase family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |