Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGATTCAGACTTTCCCTCTGCA[C/T]CAAGGACAGGTCCTCCCAGGGCTCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600755 MIM: 601915 | |||||||||||||||||||||||
Literature Links: |
SYN2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SYN2 - synapsin II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003178.5 | 938 | Intron | NP_003169.2 | |||
NM_133625.4 | 938 | Intron | NP_598328.1 | |||
XM_006713311.3 | 938 | Intron | XP_006713374.1 | |||
XM_006713312.3 | 938 | Intron | XP_006713375.1 | |||
XM_006713313.2 | 938 | Intron | XP_006713376.1 | |||
XM_017007087.1 | 938 | Intron | XP_016862576.1 |
TIMP4 - TIMP metallopeptidase inhibitor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003256.3 | 938 | Missense Mutation | ATG,GTG | M,V 143 | NP_003247.1 |