Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGGAGCGGGTCCGGGTGCTGC[A/C]GGGTGTCCGGGTGAGCGGCGCCGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615634 | |||||||||||||||||||||||
Literature Links: |
CHCHD6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CHCHD6 - coiled-coil-helix-coiled-coil-helix domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320610.1 | 149 | Missense Mutation | CAG,CCG | Q,P 26 | NP_001307539.1 | |
NM_032343.2 | 149 | Missense Mutation | CAG,CCG | Q,P 26 | NP_115719.1 | |
XM_011513234.1 | 149 | Intron | XP_011511536.1 | |||
XM_011513235.2 | 149 | Missense Mutation | CAG,CCG | Q,P 26 | XP_011511537.1 | |
XM_017007320.1 | 149 | Missense Mutation | CAG,CCG | Q,P 26 | XP_016862809.1 | |
XM_017007321.1 | 149 | Missense Mutation | CAG,CCG | Q,P 26 | XP_016862810.1 | |
XM_017007322.1 | 149 | Missense Mutation | CAG,CCG | Q,P 26 | XP_016862811.1 |