Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACAGGCACTGATAAGAAGTACC[C/T]GCTGATGAAGCAGCGTGGGTTCTAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603332 | |||||||||||||||||||||||
Literature Links: |
DNAH1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DNAH1 - dynein axonemal heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015512.4 | 3419 | Missense Mutation | CCG,CTG | P,L 90 | NP_056327.4 | |
XM_017006129.1 | 3419 | Missense Mutation | CCG,CTG | P,L 90 | XP_016861618.1 | |
XM_017006130.1 | 3419 | Missense Mutation | CCG,CTG | P,L 90 | XP_016861619.1 | |
XM_017006131.1 | 3419 | Missense Mutation | CCG,CTG | P,L 90 | XP_016861620.1 | |
XM_017006132.1 | 3419 | Missense Mutation | CCG,CTG | P,L 90 | XP_016861621.1 | |
XM_017006133.1 | 3419 | Missense Mutation | CCG,CTG | P,L 90 | XP_016861622.1 |