Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTCACTTGCTGCGCTGTGAGGC[G/T]ACTCCCTGAGGGTATTTCTGCTCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608289 | |||||||||||||||||||||||
Literature Links: |
IGF2BP2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IGF2BP2 - insulin like growth factor 2 mRNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007225.1 | Intron | NP_001007226.1 | ||||
NM_001291869.1 | Intron | NP_001278798.1 | ||||
NM_001291872.1 | Intron | NP_001278801.1 | ||||
NM_001291873.1 | Intron | NP_001278802.1 | ||||
NM_001291874.1 | Intron | NP_001278803.1 | ||||
NM_001291875.1 | Intron | NP_001278804.1 | ||||
NM_006548.4 | Intron | NP_006539.3 | ||||
XM_006713477.3 | Intron | XP_006713540.1 | ||||
XM_006713478.3 | Intron | XP_006713541.1 | ||||
XM_017005557.1 | Intron | XP_016861046.1 | ||||
XM_017005558.1 | Intron | XP_016861047.1 | ||||
XM_017005559.1 | Intron | XP_016861048.1 | ||||
XM_017005560.1 | Intron | XP_016861049.1 | ||||
XM_017005561.1 | Intron | XP_016861050.1 | ||||
XM_017005562.1 | Intron | XP_016861051.1 | ||||
XM_017005563.1 | Intron | XP_016861052.1 | ||||
XM_017005564.1 | Intron | XP_016861053.1 |