Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCATGTGCTGCCCGGGCATCA[C/T]GGAACCTGGGCCTGGTATCTGACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601732 | ||||||||||||||||||||
Literature Links: |
SMARCC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMARCC1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003074.3 | 2916 | Missense Mutation | ATG,GTG | M,V 1023 | NP_003065.3 | |
XM_011534034.1 | 2916 | Missense Mutation | ATG,GTG | M,V 914 | XP_011532336.1 |