Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGACCTCACCTGTTCTGATGGGG[C/G]CTGCCATTTCTGCCCCTTCTGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612120 | ||||||||||||||||||||
Literature Links: |
CIDEC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIDEC - cell death inducing DFFA like effector c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199551.1 | 438 | Missense Mutation | CCC,GCC | P,A 128 | NP_001186480.1 | |
NM_001199552.1 | 438 | Missense Mutation | CCC,GCC | P,A 118 | NP_001186481.1 | |
NM_001199623.1 | 438 | Missense Mutation | CCC,GCC | P,A 131 | NP_001186552.1 | |
NM_001321142.1 | 438 | Missense Mutation | CCC,GCC | P,A 118 | NP_001308071.1 | |
NM_001321143.1 | 438 | Missense Mutation | CCC,GCC | P,A 44 | NP_001308072.1 | |
NM_001321144.1 | 438 | Missense Mutation | CCC,GCC | P,A 44 | NP_001308073.1 | |
NM_022094.3 | 438 | Missense Mutation | CCC,GCC | P,A 118 | NP_071377.2 |