Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCAAGGAGGGGTCCCTCAGAGC[A/C]CCTGCCATACCCCTGCACAGTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603065 | ||||||||||||||||||||
Literature Links: |
NR1I2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR1I2 - nuclear receptor subfamily 1 group I member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003889.3 | 96 | Intron | NP_003880.3 | |||
NM_022002.2 | 96 | Silent Mutation | GCA,GCC | A,A 16 | NP_071285.1 | |
NM_033013.2 | 96 | Intron | NP_148934.1 |