Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGCCCCCCTCAGGCCATGGCCC[C/T]GGCCACCACGGTCACTATTGAATGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611337 | |||||||||||||||||||||||
Literature Links: |
DNAJB8 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DNAJB8 - DnaJ heat shock protein family (Hsp40) member B8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153330.4 | 2935 | Missense Mutation | CAG,CGG | Q,R 126 | NP_699161.1 | |
XM_006713519.3 | 2935 | Missense Mutation | CAG,CGG | Q,R 126 | XP_006713582.1 |
DNAJB8-AS1 - DNAJB8 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |