Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACTCGCATCTGAAGGGCTTCTC[G/T]CCTGGCGGAGGCAACGCAGAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608948 | ||||||||||||||||||||
Literature Links: |
ZIC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZIC4 - Zic family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168378.1 | 1152 | Silent Mutation | GGA,GGC | G,G 280 | NP_001161850.1 | |
NM_001168379.1 | 1152 | Silent Mutation | GGA,GGC | G,G 268 | NP_001161851.1 | |
NM_001243256.1 | 1152 | Missense Mutation | AGA,AGC | R,S 24 | NP_001230185.1 | |
NM_032153.5 | 1152 | Silent Mutation | GGA,GGC | G,G 230 | NP_115529.2 | |
XM_017007314.1 | 1152 | Intron | XP_016862803.1 |