Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACATCGTGCCTGCTCCGGGCCCC[A/G]TGCTGGCGCCTACGCACCTGCAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610925 MIM: 614995 | ||||||||||||||||||||
Literature Links: |
IL17RC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL17RC - interleukin 17 receptor C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203263.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | NP_001190192.1 | |
NM_001203264.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | NP_001190193.1 | |
NM_001203265.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | NP_001190194.1 | |
NM_032732.5 | 420 | Missense Mutation | ATG,GTG | M,V 60 | NP_116121.2 | |
NM_153460.3 | 420 | Missense Mutation | ATG,GTG | M,V 60 | NP_703190.1 | |
NM_153461.3 | 420 | Missense Mutation | ATG,GTG | M,V 131 | NP_703191.1 | |
XM_017007326.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862815.1 | |
XM_017007327.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862816.1 | |
XM_017007328.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862817.1 | |
XM_017007329.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862818.1 | |
XM_017007330.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862819.1 | |
XM_017007331.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862820.1 | |
XM_017007332.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862821.1 | |
XM_017007333.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862822.1 | |
XM_017007334.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862823.1 | |
XM_017007335.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862824.1 | |
XM_017007336.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862825.1 | |
XM_017007337.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862826.1 | |
XM_017007338.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862827.1 | |
XM_017007339.1 | 420 | Missense Mutation | ATG,GTG | M,V 131 | XP_016862828.1 | |
XM_017007340.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862829.1 | |
XM_017007341.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862830.1 | |
XM_017007342.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862831.1 | |
XM_017007343.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862832.1 | |
XM_017007344.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862833.1 | |
XM_017007345.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862834.1 | |
XM_017007346.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862835.1 | |
XM_017007347.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862836.1 | |
XM_017007348.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862837.1 | |
XM_017007349.1 | 420 | Missense Mutation | ATG,GTG | M,V 60 | XP_016862838.1 |
IL17RE - interleukin 17 receptor E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |