Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGGGTAGGCTCCTCGGGGCTC[C/T]GGGCCTCACGCAGGAAGCGCTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606994 | ||||||||||||||||||||
Literature Links: |
LOC101929697 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929697 - uncharacterized LOC101929697 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNK2 - tyrosine kinase non receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010938.1 | 3182 | Missense Mutation | CAG,CGG | Q,R 958 | NP_001010938.1 | |
NM_001308046.1 | 3182 | Missense Mutation | CAG,CGG | Q,R 912 | NP_001294975.1 | |
NM_005781.4 | 3182 | Missense Mutation | CAG,CGG | Q,R 880 | NP_005772.3 | |
XM_005269268.3 | 3182 | Missense Mutation | CAG,CGG | Q,R 958 | XP_005269325.1 | |
XM_005269270.3 | 3182 | Missense Mutation | CAG,CGG | Q,R 895 | XP_005269327.1 | |
XM_005269274.3 | 3182 | Missense Mutation | CAG,CGG | Q,R 656 | XP_005269331.1 | |
XM_005269275.3 | 3182 | Missense Mutation | CAG,CGG | Q,R 581 | XP_005269332.1 | |
XM_011512317.2 | 3182 | Missense Mutation | CAG,CGG | Q,R 1059 | XP_011510619.1 | |
XM_011512318.2 | 3182 | Missense Mutation | CAG,CGG | Q,R 927 | XP_011510620.2 | |
XM_011512320.1 | 3182 | Missense Mutation | CAG,CGG | Q,R 895 | XP_011510622.1 | |
XM_011512321.2 | 3182 | Missense Mutation | CAG,CGG | Q,R 819 | XP_011510623.1 | |
XM_017005508.1 | 3182 | Missense Mutation | CAG,CGG | Q,R 912 | XP_016860997.1 | |
XM_017005509.1 | 3182 | Missense Mutation | CAG,CGG | Q,R 912 | XP_016860998.1 | |
XM_017005510.1 | 3182 | Missense Mutation | CAG,CGG | Q,R 927 | XP_016860999.1 |