Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTGACACACGTACCATTCGCT[C/T]AAGTCGGCGGTACGCGCCGCCACCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608948 | |||||||||||||||||||||||
Literature Links: |
ZIC4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZIC4 - Zic family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168378.1 | 1458 | Silent Mutation | TTA,TTG | L,L 382 | NP_001161850.1 | |
NM_001168379.1 | 1458 | Silent Mutation | TTA,TTG | L,L 370 | NP_001161851.1 | |
NM_001243256.1 | 1458 | Silent Mutation | TTA,TTG | L,L 126 | NP_001230185.1 | |
NM_032153.5 | 1458 | Silent Mutation | TTA,TTG | L,L 332 | NP_115529.2 | |
XM_017007314.1 | 1458 | Intron | XP_016862803.1 |