Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTACAGCTACACTCCTAGGAA[A/G]GACCACCATCAATGATTACCAGGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 155970 | |||||||||||||||||||||||
Literature Links: |
CD200 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CD200 - CD200 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004196.3 | 505 | Missense Mutation | AAG,AGG | K,R 22 | NP_001004196.2 | |
NM_001318826.1 | 505 | Intron | NP_001305755.1 | |||
NM_001318828.1 | 505 | Intron | NP_001305757.1 | |||
NM_001318830.1 | 505 | Intron | NP_001305759.1 | |||
NM_005944.6 | 505 | Intron | NP_005935.4 | |||
XM_005247481.4 | 505 | Missense Mutation | AAG,AGG | K,R 22 | XP_005247538.1 | |
XM_005247482.2 | 505 | Intron | XP_005247539.1 | |||
XM_005247485.2 | 505 | Intron | XP_005247542.1 | |||
XM_005247486.2 | 505 | Intron | XP_005247543.1 | |||
XM_005247488.2 | 505 | Intron | XP_005247545.1 | |||
XM_017006452.1 | 505 | UTR 5 | XP_016861941.1 | |||
XM_017006453.1 | 505 | Intron | XP_016861942.1 | |||
XM_017006454.1 | 505 | UTR 5 | XP_016861943.1 | |||
XM_017006455.1 | 505 | Intron | XP_016861944.1 | |||
XM_017006456.1 | 505 | UTR 5 | XP_016861945.1 | |||
XM_017006457.1 | 505 | Intron | XP_016861946.1 | |||
XM_017006458.1 | 505 | UTR 5 | XP_016861947.1 | |||
XM_017006459.1 | 505 | Intron | XP_016861948.1 |
LOC105374042 - uncharacterized LOC105374042 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |