Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGAAACACCCTGGTGCTCGGTT[C/T]CTCGGGGAGCTCTGGATGGAGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF662 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF662 - zinc finger protein 662 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134656.1 | 472 | Missense Mutation | CCT,TCT | P,S 96 | NP_001128128.1 | |
NM_207404.3 | 472 | Missense Mutation | CCT,TCT | P,S 36 | NP_997287.2 |