Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCGACGGCAGTGGCTGCAGCGG[A/G]CACGGCCGAAGGACCGGGAGGCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602303 | ||||||||||||||||||||
Literature Links: |
KAT2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KAT2B - lysine acetyltransferase 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003884.4 | 256 | Missense Mutation | GAC,GGC | D,G 62 | NP_003875.3 | |
XM_005265528.4 | 256 | Missense Mutation | GAC,GGC | D,G 62 | XP_005265585.1 | |
XM_017007423.1 | 256 | Intron | XP_016862912.1 | |||
XM_017007424.1 | 256 | Intron | XP_016862913.1 |