Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGGTTGCTTTAATTTTTCCCA[A/T]ACCTGGCTTGAGGGAAAATTCACAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ERICH6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ERICH6 - glutamate rich 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308234.1 | 2299 | Silent Mutation | GTA,GTT | V,V 475 | NP_001295163.1 | |
NM_152394.4 | 2299 | Silent Mutation | GTA,GTT | V,V 621 | NP_689607.2 | |
XM_005247120.3 | 2299 | UTR 3 | XP_005247177.1 |