Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCGGTAGGTGGTTTTCTCTCC[C/G]ACAGGCACCGCCCGGACGGTGGCAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 155750 | |||||||||||||||||||||||
Literature Links: |
MELTF PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MELTF - melanotransferrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005929.5 | 2240 | Silent Mutation | GTC,GTG | V,V 683 | NP_005920.2 | |
NM_033316.3 | 2240 | Intron | NP_201573.1 | |||
XM_006713643.3 | 2240 | Silent Mutation | GTC,GTG | V,V 709 | XP_006713706.2 | |
XM_011512850.2 | 2240 | Silent Mutation | GTC,GTG | V,V 710 | XP_011511152.1 | |
XM_011512851.2 | 2240 | Intron | XP_011511153.1 |
MFI2-AS1 - MFI2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |