Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCCCTGACCTTCAAGGAAGAT[A/T]TGAGCTATCTGGGAAAAACAGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613250 MIM: 613822 | ||||||||||||||||||||
Literature Links: |
EBLN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EBLN2 - endogenous Bornavirus-like nucleoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018029.4 | 653 | Missense Mutation | TAT,TTT | Y,F 77 | NP_060499.3 |
PPP4R2 - protein phosphatase 4 regulatory subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318025.1 | 653 | Intron | NP_001304954.1 | |||
NM_001318026.1 | 653 | Intron | NP_001304955.1 | |||
NM_001318027.1 | 653 | Intron | NP_001304956.1 | |||
NM_001318028.1 | 653 | Intron | NP_001304957.1 | |||
NM_174907.3 | 653 | Intron | NP_777567.1 |