Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGACGAGGAGTTCCAGCCATCA[C/T]AGCTGTTCGACTTCGATGATTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604226 | ||||||||||||||||||||
Literature Links: |
ARPC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARPC4 - actin related protein 2/3 complex subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARPC4-TTLL3 - ARPC4-TTLL3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198793.1 | 1109 | Nonsense Mutation | CAG,TAG | Q,* 179 | NP_001185722.1 |
TTLL3 - tubulin tyrosine ligase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025930.3 | 1109 | Nonsense Mutation | CAG,TAG | Q,* 228 | NP_001021100.3 |