Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGGCCCTGGCTGGTGGGTCAC[A/G]CCGTGGATGGCTGGGAGCAGCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601253 MIM: 167055 | ||||||||||||||||||||
Literature Links: |
CAV3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAV3 - caveolin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OXTR - oxytocin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000916.3 | 1764 | Missense Mutation | GCG,GTG | A,V 389 | NP_000907.2 | |
XM_011533762.2 | 1764 | Missense Mutation | GCG,GTG | A,V 389 | XP_011532064.1 |
SSUH2 - ssu-2 homolog (C. elegans) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |