Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATAGCGTCTGCGGATGCGCCACA[G/T]GCGGGCCTGGGACTGCAGTGTGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IQCF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQCF1 - IQ motif containing F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152397.2 | 507 | Missense Mutation | ATG,CTG | M,L 137 | NP_689610.2 | |
XM_011533366.2 | 507 | Missense Mutation | ATG,CTG | M,L 102 | XP_011531668.1 | |
XM_017005729.1 | 507 | Missense Mutation | ATG,CTG | M,L 102 | XP_016861218.1 |