Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGAAGGTAACTTTGTGCAGCAT[C/T]TACCATTTTCCAGGAATTGTAGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606279 MIM: 602498 | ||||||||||||||||||||
Literature Links: |
ABI3BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABI3BP - ABI family member 3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015429.3 | 5335 | Silent Mutation | TAA,TAG | *,* 1076 | NP_056244.2 | |
XM_005247281.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1794 | XP_005247338.1 | |
XM_005247282.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1780 | XP_005247339.1 | |
XM_005247283.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1778 | XP_005247340.1 | |
XM_005247284.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1775 | XP_005247341.1 | |
XM_005247285.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1774 | XP_005247342.1 | |
XM_005247287.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1772 | XP_005247344.1 | |
XM_005247288.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1770 | XP_005247345.1 | |
XM_005247289.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1769 | XP_005247346.1 | |
XM_005247290.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1769 | XP_005247347.1 | |
XM_005247291.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1768 | XP_005247348.1 | |
XM_005247293.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1758 | XP_005247350.1 | |
XM_005247294.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1755 | XP_005247351.1 | |
XM_005247295.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1748 | XP_005247352.1 | |
XM_005247296.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1747 | XP_005247353.1 | |
XM_005247298.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1746 | XP_005247355.1 | |
XM_005247299.3 | 5335 | Silent Mutation | TAA,TAG | *,* 1745 | XP_005247356.1 | |
XM_005247300.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1736 | XP_005247357.1 | |
XM_005247301.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1727 | XP_005247358.1 | |
XM_005247302.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1726 | XP_005247359.1 | |
XM_005247303.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1707 | XP_005247360.1 | |
XM_005247305.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1679 | XP_005247362.1 | |
XM_005247306.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1658 | XP_005247363.1 | |
XM_005247309.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1496 | XP_005247366.1 | |
XM_006713569.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1743 | XP_006713632.1 | |
XM_006713570.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1732 | XP_006713633.1 | |
XM_006713571.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1722 | XP_006713634.1 | |
XM_006713572.2 | 5335 | Silent Mutation | TAA,TAG | *,* 1720 | XP_006713635.1 | |
XM_011512639.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1774 | XP_011510941.1 | |
XM_011512640.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1772 | XP_011510942.1 | |
XM_011512641.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1769 | XP_011510943.1 | |
XM_011512643.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1767 | XP_011510945.1 | |
XM_011512644.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1766 | XP_011510946.1 | |
XM_011512645.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1766 | XP_011510947.1 | |
XM_011512646.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1763 | XP_011510948.1 | |
XM_011512647.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1749 | XP_011510949.1 | |
XM_011512648.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1748 | XP_011510950.1 | |
XM_011512649.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1746 | XP_011510951.1 | |
XM_011512651.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1727 | XP_011510953.1 | |
XM_011512652.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1723 | XP_011510954.1 | |
XM_011512653.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1720 | XP_011510955.1 | |
XM_011512654.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1717 | XP_011510956.1 | |
XM_011512656.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1695 | XP_011510958.1 | |
XM_011512657.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1694 | XP_011510959.1 | |
XM_011512658.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1690 | XP_011510960.1 | |
XM_011512659.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1683 | XP_011510961.1 | |
XM_011512660.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1673 | XP_011510962.1 | |
XM_017006105.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1775 | XP_016861594.1 | |
XM_017006106.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1769 | XP_016861595.1 | |
XM_017006107.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1725 | XP_016861596.1 | |
XM_017006108.1 | 5335 | Silent Mutation | TAA,TAG | *,* 1695 | XP_016861597.1 |
TFG - TRK-fused gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |