Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTAGATGGCTCTGTTGCCTCT[C/T]ACGCCAGTTCTTTTGGCAGTTGCAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605709 MIM: 616112 | |||||||||||||||||||||||
Literature Links: |
ARL6IP5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARL6IP5 - ADP ribosylation factor like GTPase 6 interacting protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMOD3 - leiomodin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304418.1 | 1891 | Silent Mutation | TAA,TGA | *,* 561 | NP_001291347.1 | |
NM_198271.4 | 1891 | Silent Mutation | TAA,TGA | *,* 561 | NP_938012.2 |