Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCACACTGTTTAGTCTTTTGAA[C/T]GGCTCTTCCTTCTCAACGAGAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601854 | ||||||||||||||||||||
Literature Links: |
DGKG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGKG - diacylglycerol kinase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080744.1 | 2783 | Missense Mutation | CAT,CGT | H,R 763 | NP_001074213.1 | |
NM_001080745.1 | 2783 | Missense Mutation | CAT,CGT | H,R 749 | NP_001074214.1 | |
NM_001346.2 | 2783 | Missense Mutation | CAT,CGT | H,R 788 | NP_001337.2 |