Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACAGCAGAAGCTCTTGTGGAAG[C/G]TGGGAAAGCCCCTAAGTTGACTCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603081 | |||||||||||||||||||||||
Literature Links: |
ADPRH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ADPRH - ADP-ribosylarginine hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001125.3 | 816 | Missense Mutation | GCT,GGT | A,G 70 | NP_001116.1 | |
NM_001291949.1 | 816 | Missense Mutation | GCT,GGT | A,G 70 | NP_001278878.1 | |
NM_001291950.1 | 816 | Intron | NP_001278879.1 | |||
XM_011512438.2 | 816 | Missense Mutation | GCT,GGT | A,G 70 | XP_011510740.1 |